Iron(II) tetrafluoroborate hexahydrate 95%

Supplier: Ambeed
Danger

Synonyms: Ferrous tetrafluoroborate

A775119-5G A775119-25G
76652-940EA 18 USD
76652-940 77552-392
Iron(II) tetrafluoroborate hexahydrate 95%
Iron(II) tetrafluoroborate hexahydrate

Precaución: for Research Use Only

Formula: 2(BF₄)·Fe·6H₂O
MW: 337.546 g/mol
MDL Number: MFCD00192235
CAS Number: 13877-16-2
UN: 3260
ADR: 8 Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR