(E)-1-(6-Methoxynaphthalen-2-yl)-3-(pyridin-3-yl)prop-2-en-1-one 99+%
Supplier: Ambeed
|
Warning
|
Synonyms:
DMU2139
A774966-10MG
A774966-1MG
A774966-25MG
A774966-50MG
A774966-5MG
76652-930EA
18.04
USD
76652-930
76652-932
76652-934
76652-936
76652-938
(E)-1-(6-Methoxynaphthalen-2-yl)-3-(pyridin-3-yl)prop-2-en-1-one 99+%
(E)-1-(6-Methoxynaphthalen-2-yl)-3-(pyridin-3-yl)prop-2-en-1-one
Precaución: for Research Use Only
Formula:
C₁₉H₁₅NO₂ MW: 289.33 g/mol |
CAS Number:
1821143-80-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...