2,4-Hexadien-1-ol 98% stabilized

Supplier: Ambeed
Danger

Synonyms: Sorbic Alcohol

Stabilization: Stabilized with Tocopherols 0.1%

A774396-1G A774396-25G A774396-5G
76652-922EA 23.34 USD
76652-922 76652-924 76652-926
2,4-Hexadien-1-ol 98% stabilized
2,4-Hexadien-1-ol

Precaución: for Research Use Only

Formula: C₆H₁₀O
MW: 98.15 g/mol
MDL Number: MFCD00002925
CAS Number: 111-28-4
UN: 1759
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR