5-Bromo-N-((4,6-dimethyl-2-oxo-1,2-dihydropyridin-3-yl)methyl)-3-(ethyl(tetrahydro-2H-pyran-4-yl)amino)-2-methylbenzamide 98% (contains 2-3%DCM)

Supplier: Ambeed
Warning

A773420-100MG A773420-250MG
76652-910EA 83 USD
76652-910 77573-486
5-Bromo-N-((4,6-dimethyl-2-oxo-1,2-dihydropyridin-3-yl)methyl)-3-(ethyl(tetrahydro-2H-pyran-4-yl)amino)-2-methylbenzamide 98% (contains 2-3%DCM)
5-Bromo-N-((4,6-dimethyl-2-oxo-1,2-dihydropyridin-3-yl)methyl)-3-(ethyl(tetrahydro-2H-pyran-4-yl)amino)-2-methylbenzamide

Precaución: for Research Use Only

Formula: C₂₃H₃₀BrN₃O₃
MDL Number: MFCD28167870
CAS Number: 1403257-80-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR