(1-(Morpholinomethyl)cyclopropyl)methanol 95%

Supplier: Ambeed
Warning

A772361-1G A772361-250MG A772361-5G
76652-904EA 97 USD
76652-904 76652-906 76652-908
(1-(Morpholinomethyl)cyclopropyl)methanol 95%
(1-(Morpholinomethyl)cyclopropyl)methanol

Precaución: for Research Use Only

Formula: C₉H₁₇NO₂
MDL Number: MFCD19313522
CAS Number: 1267956-65-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR