3-Nitro-2-(trifluoromethyl)benzonitrile 97%

Supplier: Ambeed
Warning

A771611-100MG A771611-250MG
76652-898EA 18 USD
76652-898 76652-900
3-Nitro-2-(trifluoromethyl)benzonitrile 97%
3-Nitro-2-(trifluoromethyl)benzonitrile

Precaución: for Research Use Only

Formula: C₈H₃F₃N₂O₂
MDL Number: MFCD16606331
CAS Number: 1227516-56-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR