Triethylene Glycol Mono(2-propynyl) ether 95%

Supplier: Ambeed
Warning

Synonyms: Propargyl-PEG3-alcohol, 2-[2-[2-(2-Propynyloxy)ethoxy]ethoxy]ethanol, Triethylene Glycol Monopropargyl ether

A771417-1G A771417-250MG A771417-5G A771417-100MG A771417-25G
76652-892EA 28 USD
76652-892 76652-894 77569-464 77618-116 77618-118
Triethylene Glycol Mono(2-propynyl) ether 95%
Triethylene Glycol Mono(2-propynyl) ether

Precaución: For research use only.

Formula: C₉H₁₆O₄
MW: 188.22 g/mol
MDL Number: MFCD13189984
CAS Number: 208827-90-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR