2,4,6-Tribromobenzene-1,3,5-triol 95%

Supplier: Ambeed
Warning

A771030-1G A771030-5G
76652-882EA 119 USD
76652-882 76652-884
2,4,6-Tribromobenzene-1,3,5-triol 95%
2,4,6-Tribromobenzene-1,3,5-triol

Precaución: for Research Use Only

Formula: C₆H₃Br₃O₃
MDL Number: MFCD01706660
CAS Number: 3354-82-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR