1,2,4-Trimethoxybenzene 95%

Supplier: Ambeed
Warning

A767549-100G A767549-25G A767549-500G A767549-5G
76652-864EA 51 USD
76652-864 76652-866 76652-868 76652-870
1,2,4-Trimethoxybenzene 95%
1,2,4-Trimethoxybenzene

Precaución: for Research Use Only

Formula: C₉H₁₂O₃
MDL Number: MFCD00008360
CAS Number: 135-77-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR