1-(2-Methoxyethyl)-1-methylpyrrolidinium bis(fluorosulfonyl)imide 95%

Supplier: Ambeed
Danger

A765610-1G A765610-250MG A765610-5G
76652-850EA 23 USD
76652-850 76652-852 76652-854
1-(2-Methoxyethyl)-1-methylpyrrolidinium bis(fluorosulfonyl)imide 95%
1-(2-Methoxyethyl)-1-methylpyrrolidinium bis(fluorosulfonyl)imide

Precaución: for Research Use Only

Formula: C₈H₁₈F₂N₂O₅S₂
MDL Number: MFCD30749293
CAS Number: 1235234-47-5
UN: 1760
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR