tert-Butyl (2-(2-(2-(prop-2-yn-1-yloxy)ethoxy)ethoxy)ethyl)carbamate 95%

Supplier: Ambeed
Warning

Synonyms: Boc-NH-PEG3-propargyl

A765565-100MG A765565-1G A765565-250MG
76652-844EA 39.26 USD
76652-844 76652-846 76652-848
tert-Butyl (2-(2-(2-(prop-2-yn-1-yloxy)ethoxy)ethoxy)ethyl)carbamate 95%
tert-Butyl (2-(2-(2-(prop-2-yn-1-yloxy)ethoxy)ethoxy)ethyl)carbamate

Precaución: for Research Use Only

Formula: C₁₄H₂₅NO₅
MW: 287.3520 g/mol
MDL Number: MFCD30534430
CAS Number: 1333880-60-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR