Boc-4-hydroxy-D-phenylglycine 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
(R)-2-((tert-Butoxycarbonyl)amino)-2-(4-hydroxyphenyl)acetic acid, Boc-D-Phg(4-OH)-OH, Boc-D-4-OH-Phg-OH
A765520-1G
A765520-250MG
A765520-5G
76652-838EA
14.86
USD
76652-838
76652-840
76652-842
Boc-4-hydroxy-D-phenylglycine 98%
Boc-4-hydroxy-D-phenylglycine
Precaución: for Research Use Only
Formula:
C₁₃H₁₇NO₅ MW: 267.28 g/mol |
MDL Number:
MFCD01861312 CAS Number: 27460-85-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...