tert-Butyl (20-hydroxy-3,6,9,12,15,18-hexaoxaicosyl)carbamate 95%

Supplier: Ambeed
Warning

Synonyms: N-Boc-PEG7-alcohol

A765121-100MG A765121-250MG A765121-5G A765121-1G
76652-832EA 63.66 USD
76652-832 76652-834 76652-836 77552-354
tert-Butyl (20-hydroxy-3,6,9,12,15,18-hexaoxaicosyl)carbamate 95%
tert-Butyl (20-hydroxy-3,6,9,12,15,18-hexaoxaicosyl)carbamate

Precaución: for Research Use Only

Formula: C₁₉H₃₉NO₉
MW: 425.5143 g/mol
MDL Number: MFCD29049415
CAS Number: 1292268-13-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR