tert-Butyl 17-bromoheptadecanoate 98%

Supplier: Ambeed
Warning

A764942-1G A764942-5G
76652-830EA 101.85 USD
76652-830 77599-056
tert-Butyl 17-bromoheptadecanoate 98%
tert-Butyl 17-bromoheptadecanoate

Precaución: for Research Use Only

Formula: C₂₁H₄₁BrO₂
MW: 405.4531 g/mol
MDL Number: MFCD31728061
CAS Number: 2428400-70-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR