tert-Butyl (2-(2-(2-(2-(aminooxy)ethoxy)ethoxy)ethoxy)ethyl)carbamate 95%

Supplier: Ambeed
Warning

Synonyms: Aminooxy-PEG3-C2-NH-Boc

A764928-1G A764928-250MG A764928-100MG A764928-5G
76652-826EA 250 USD
76652-826 76652-828 77622-908 77783-994
tert-Butyl (2-(2-(2-(2-(aminooxy)ethoxy)ethoxy)ethoxy)ethyl)carbamate 95%
tert-Butyl (2-(2-(2-(2-(aminooxy)ethoxy)ethoxy)ethoxy)ethyl)carbamate

Precaución: for Research Use Only

Formula: C₁₃H₂₈N₂O₆
MDL Number: MFCD30536148
CAS Number: 2062663-65-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR