tert-Butyl (26-hydroxy-3,6,9,12,15,18,21,24-octaoxahexacosyl)carbamate 95%

Supplier: Ambeed
Warning

Synonyms: N-Boc-PEG9-alcohol

A764888-100MG A764888-1G A764888-250MG
76652-820EA 74 USD
76652-820 76652-822 76652-824
tert-Butyl (26-hydroxy-3,6,9,12,15,18,21,24-octaoxahexacosyl)carbamate 95%
tert-Butyl (26-hydroxy-3,6,9,12,15,18,21,24-octaoxahexacosyl)carbamate

Precaución: for Research Use Only

Formula: C₂₃H₄₇NO₁₁
MDL Number: MFCD30723250
CAS Number: 2112731-44-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR