1-Butylpyridinium bis(trifluoromethanesulfonyl)imide 98%

Supplier: Ambeed
Warning

A764579-100G A764579-1G A764579-25G A764579-5G
76652-814EA 182 USD
76652-814 76652-816 76652-818 77595-208
1-Butylpyridinium bis(trifluoromethanesulfonyl)imide 98%
1-Butylpyridinium bis(trifluoromethanesulfonyl)imide

Precaución: for Research Use Only

Formula: C₁₁H₁₄F₆N₂O₄S₂
MW: 416.35 g/mol
MDL Number: MFCD16038143
CAS Number: 187863-42-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR