Potassium (Acetoxymethyl)trifluoroborate 98%

Supplier: Ambeed
Warning

Synonyms: Potassium (acetoxymethyl)trifluoroborate

A763917-100MG A763917-1G A763917-5G
76652-812EA 48.81 USD
76652-812 77552-346 77622-906
Potassium (Acetoxymethyl)trifluoroborate 98%
Potassium (Acetoxymethyl)trifluoroborate

Precaución: for Research Use Only

Formula: C₃H₅BF₃KO₂
MW: 179.97 g/mol
MDL Number: MFCD28023894
CAS Number: 910251-35-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR