2-(4-(Thien-2-yl)phenyl)thiophene 93%

Supplier: Ambeed
Warning

A763724-100MG A763724-1G A763724-250MG
76652-806EA 47.75 USD
76652-806 76652-808 76652-810
2-(4-(Thien-2-yl)phenyl)thiophene 93%
2-(4-(Thien-2-yl)phenyl)thiophene

Precaución: for Research Use Only

Formula: C₁₄H₁₀S₂
MW: 242.3592 g/mol
MDL Number: MFCD11224849
CAS Number: 23354-94-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR