11-([1,1'-Biphenyl]-3-yl)-11,12-dihydroindolo[2,3-a]carbazole 97%

Supplier: Ambeed
Warning

A763211-250MG A763211-1G
76652-794EA 83 USD
76652-794 77622-902
11-([1,1'-Biphenyl]-3-yl)-11,12-dihydroindolo[2,3-a]carbazole 97%
11-([1,1'-Biphenyl]-3-yl)-11,12-dihydroindolo[2,3-a]carbazole

Precaución: for Research Use Only

Formula: C₃₀H₂₀N₂
CAS Number: 1449754-80-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR