4-((17S,20S)-1-(2,5-Dioxo-2,5-dihydro-1H-pyrrol-1-yl)-17-isopropyl-15,18-dioxo-20-(3-ureidopropyl)-3,6,9,12-tetraoxa-16,19-diazahenicosanamido)benzyl (4-nitrophenyl) carbonate 95%

Supplier: Ambeed
Warning

Synonyms: Mal-PEG4-Val-Cit-PAB-PNP

A763179-25MG A763179-50MG A763179-10MG A763179-100MG
76652-790EA 298.01 USD
76652-790 76652-792 77622-900 77783-985
4-((17S,20S)-1-(2,5-Dioxo-2,5-dihydro-1H-pyrrol-1-yl)-17-isopropyl-15,18-dioxo-20-(3-ureidopropyl)-3,6,9,12-tetraoxa-16,19-diazahenicosanamido)benzyl (4-nitrophenyl) carbonate 95%
4-((17S,20S)-1-(2,5-Dioxo-2,5-dihydro-1H-pyrrol-1-yl)-17-isopropyl-15,18-dioxo-20-(3-ureidopropyl)-3,6,9,12-tetraoxa-16,19-diazahenicosanamido)benzyl (4-nitrophenyl) carbonate

Precaución: for Research Use Only

Formula: C₄₀H₅₃N₇O₁₅
MDL Number: MFCD30828694
CAS Number: 2112738-09-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR