3-(2-(3-(tert-Butoxy)-3-oxopropoxy)ethoxy)propanoic acid 95%

Supplier: Ambeed
Warning

Synonyms: Acid-PEG2-C2-Boc

A763155-100MG A763155-250MG A763155-50MG A763155-1G A763155-5G
76652-784EA 86 USD
76652-784 76652-786 76652-788 77599-044 77622-898
3-(2-(3-(tert-Butoxy)-3-oxopropoxy)ethoxy)propanoic acid 95%
3-(2-(3-(tert-Butoxy)-3-oxopropoxy)ethoxy)propanoic acid

Precaución: for Research Use Only

Formula: C₁₂H₂₂O₆
MDL Number: MFCD30723272
CAS Number: 2086688-99-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR