Glycine 7-amido-4-methylcoumarin hydrochloride 95%

Supplier: Ambeed
Warning

Synonyms: Gly-AMC HCl

A762903-10MG A762903-1MG A762903-25MG A762903-50MG A762903-5MG
76652-772EA 42.44 USD
76652-772 76652-774 76652-776 76652-778 76652-780
Glycine 7-amido-4-methylcoumarin hydrochloride 95%
Glycine 7-amido-4-methylcoumarin hydrochloride

Precaución: for Research Use Only

Formula: C₁₂H₁₃ClN₂O₃
MW: 268.6962 g/mol
MDL Number: MFCD26793620
CAS Number: 208459-17-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR