tert-Butyl 3,6,9,12,15,18,21-heptaoxatetracos-23-yn-1-ylcarbamate 95%

Supplier: Ambeed
Warning

Synonyms: Boc-NH-PEG7-propargyl

A762708-100MG A762708-1G A762708-250MG
76652-766EA 61.54 USD
76652-766 76652-768 76652-770
tert-Butyl 3,6,9,12,15,18,21-heptaoxatetracos-23-yn-1-ylcarbamate 95%
tert-Butyl 3,6,9,12,15,18,21-heptaoxatetracos-23-yn-1-ylcarbamate

Precaución: for Research Use Only

Formula: C₂₂H₄₁NO₉
MW: 463.5623 g/mol
MDL Number: MFCD30828698
CAS Number: 2112737-90-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR