3-(3-(tert-Butoxy)-3-oxopropoxy)propanoic acid 95%

Supplier: Ambeed
Warning

Synonyms: Acid-PEG1-C2-Boc

A762531-100MG A762531-250MG A762531-1G
76652-762EA 59.42 USD
76652-762 76652-764 77569-450
3-(3-(tert-Butoxy)-3-oxopropoxy)propanoic acid 95%
3-(3-(tert-Butoxy)-3-oxopropoxy)propanoic acid

Precaución: For research use only.

Formula: C₁₀H₁₈O₅
MW: 218.2469 g/mol
MDL Number: MFCD30723273
CAS Number: 2086689-08-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR