(E/Z)-Geranylacetone 97% (mixture of isomers)

Supplier: Ambeed
Warning

Synonyms: 6,10-Dimethyl-5,9-undecadien-2-one, 6,10-Dimethyl-5,9-undecadien-2-one, mixture of isomers, Geranyl-2-propanone

A761725-1G A761725-25G A761725-5G A761725-100G
76652-744EA 5.31 USD
76652-744 76652-746 76652-748 77622-886
(E/Z)-Geranylacetone 97% (mixture of isomers)
(E/Z)-Geranylacetone

Precaución: for Research Use Only

Formula: (CH₃)₂C=CHCH₂CH₂C(CH₃)=CHCH₂CH₂C(O)CH₃
MW: 194.32 g/mol
MDL Number: MFCD00008910
CAS Number: 689-67-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR