5-Bromopyridin-2(1H)-one 98%

Supplier: Ambeed
Warning

A761566-10G A761566-25G A761566-5G A761566-100G
76652-732EA 11 USD
76652-732 76652-734 76652-736 77749-520
5-Bromopyridin-2(1H)-one 98%
5-Bromopyridin-2(1H)-one

Precaución: for Research Use Only

Formula: C₅H₄BrNO
MDL Number: MFCD00456288
CAS Number: 49843-23-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR