1-(9H-Fluoren-9-yl)-3-oxo-2,7,10,13,16,19,22-heptaoxa-4-azatetracosan-24-oic acid 95%

Supplier: Ambeed
Warning

Synonyms: Fmoc-NH-PEG6-CH2COOH

A761108-100MG A761108-250MG
76652-728EA 81.69 USD
76652-728 76652-730
1-(9H-Fluoren-9-yl)-3-oxo-2,7,10,13,16,19,22-heptaoxa-4-azatetracosan-24-oic acid 95%
1-(9H-Fluoren-9-yl)-3-oxo-2,7,10,13,16,19,22-heptaoxa-4-azatetracosan-24-oic acid

Precaución: for Research Use Only

Formula: C₂₉H₃₉NO₁₀
MW: 561.6207 g/mol
MDL Number: MFCD26142983
CAS Number: 437655-96-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR