2-(2-(2-(2-Aminoethoxy)ethoxy)ethoxy)acetic acid 95%

Supplier: Ambeed
Warning

Synonyms: Amino-PEG3-CH2COOH

A760737-1G A760737-100MG A760737-250MG A760737-5G
76652-726EA 85 USD
76652-726 77622-862 77622-864 77622-866
2-(2-(2-(2-Aminoethoxy)ethoxy)ethoxy)acetic acid 95%
2-(2-(2-(2-Aminoethoxy)ethoxy)ethoxy)acetic acid

Precaución: for Research Use Only

Formula: C₈H₁₇NO₅
MDL Number: MFCD24479841
CAS Number: 134978-99-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR