Methyl 7-methyl-1H-indole-6-carboxylate 96%

Supplier: Ambeed
Warning

A760270-100MG A760270-250MG A760270-1G
76652-722EA 95.49 USD
76652-722 77749-517 77749-516
Methyl 7-methyl-1H-indole-6-carboxylate 96%
Methyl 7-methyl-1H-indole-6-carboxylate

Precaución: for Research Use Only

Formula: C₁₁H₁₁NO₂
MW: 189.2100 g/mol
MDL Number: MFCD20684609
CAS Number: 733035-34-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR