2,5-Dihydroxybenzoic acid butylamine salt 98%

Supplier: Ambeed
Warning

A760253-1G A760253-250MG A760253-5G
76652-716EA 180 USD
76652-716 76652-718 76652-720
2,5-Dihydroxybenzoic acid butylamine salt 98%
2,5-Dihydroxybenzoic acid butylamine salt

Precaución: for Research Use Only

Formula: C₇H₆O₄·C₄H₁₁N
MDL Number: MFCD30725754
CAS Number: 666174-80-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR