((4-((2-Chloro-4-nitrophenyl)diazenyl)-3-propionamidophenyl)azanediyl)bis(ethane-2,1-diyl) diacetate 97%

Supplier: Ambeed
Warning

A760244-100MG A760244-1G A760244-250MG
76652-710EA 214 USD
76652-710 76652-712 76652-714
((4-((2-Chloro-4-nitrophenyl)diazenyl)-3-propionamidophenyl)azanediyl)bis(ethane-2,1-diyl) diacetate 97%
((4-((2-Chloro-4-nitrophenyl)diazenyl)-3-propionamidophenyl)azanediyl)bis(ethane-2,1-diyl) diacetate

Precaución: for Research Use Only

Formula: C₂₃H₂₆ClN₅O₇
MDL Number: MFCD30719570
CAS Number: 26850-12-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR