Hydroxy-PEG9-t-butyl ester 95%

Supplier: Ambeed
Warning

Synonyms: Hydroxy-PEG9-Boc

A760000-1G A760000-250MG
76652-698EA 103 USD
76652-698 76652-700
Hydroxy-PEG9-t-butyl ester 95%
Hydroxy-PEG9-t-butyl ester

Precaución: for Research Use Only

Formula: C₂₅H₅₀O₁₂
MDL Number: MFCD30182029
CAS Number: 2055042-66-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR