4-Fluoro-5-iodo-2-methylbenzoic acid 95%

Supplier: Ambeed
Warning

A759995-100MG A759995-1G A759995-250MG
76652-692EA 39 USD
76652-692 76652-694 76652-696
4-Fluoro-5-iodo-2-methylbenzoic acid 95%
4-Fluoro-5-iodo-2-methylbenzoic acid

Precaución: for Research Use Only

Formula: C₈H₆FIO₂
MDL Number: MFCD26863042
CAS Number: 1533441-18-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR