1-[(Trimethylsilyl)ethynyl]-1,2-benziodoxol-3(1H)-one 98%

Supplier: Ambeed
Warning

A759880-1G A759880-250MG A759880-5G
76652-686EA 81 USD
76652-686 76652-688 76652-690
1-[(Trimethylsilyl)ethynyl]-1,2-benziodoxol-3(1H)-one 98%
1-[(Trimethylsilyl)ethynyl]-1,2-benziodoxol-3(1H)-one

Precaución: for Research Use Only

Formula: C₁₂H₁₃IO₂Si
MDL Number: MFCD29765750
CAS Number: 181934-29-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR