2-CHloro-5-iodo-4-methyl-benzoic acid methyl ester 95%
Supplier: Ambeed
|
Warning
|
A759717-1G
A759717-250MG
76652-676EA
23
USD
76652-676
76652-678
2-CHloro-5-iodo-4-methyl-benzoic acid methyl ester 95%
2-CHloro-5-iodo-4-methyl-benzoic acid methyl ester
Precaución: for Research Use Only
Formula:
C₉H₈ClIO₂ |
MDL Number:
MFCD30162939 CAS Number: 1699509-67-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...