2-(2-(2-(5-((3aS,4S,6aR)-2-Oxohexahydro-1H-thieno[3,4-d]imidazol-4-yl)pentanamido)ethoxy)ethoxy)acetic acid 95%

Supplier: Ambeed
Warning

A759615-1G A759615-100MG A759615-250MG
76652-672EA 358.59 USD
76652-672 77622-846 77622-848
2-(2-(2-(5-((3aS,4S,6aR)-2-Oxohexahydro-1H-thieno[3,4-d]imidazol-4-yl)pentanamido)ethoxy)ethoxy)acetic acid 95%
2-(2-(2-(5-((3aS,4S,6aR)-2-Oxohexahydro-1H-thieno[3,4-d]imidazol-4-yl)pentanamido)ethoxy)ethoxy)acetic acid

Precaución: for Research Use Only

Formula: C₁₆H₂₇N₃O₆S
MW: 389.4671 g/mol
MDL Number: MFCD13185030
CAS Number: 1238575-77-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR