(5-Bromo-4-fluoro-2-nitro-phenyl)-methyl-amine 95%

Supplier: Ambeed
Danger

A759466-1G A759466-250MG
76652-668EA 275.84 USD
76652-668 76652-670
(5-Bromo-4-fluoro-2-nitro-phenyl)-methyl-amine 95%
(5-Bromo-4-fluoro-2-nitro-phenyl)-methyl-amine

Precaución: for Research Use Only

Formula: C₇H₆BrFN₂O₂
MW: 249.04 g/mol
MDL Number: MFCD30073170
CAS Number: 1356483-97-0
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR