Sodium bis(fluorosulfonyl)imide 95%

Supplier: Ambeed
Danger

A759439-1G A759439-25G A759439-5G
76652-662EA 15 USD
76652-662 76652-664 76652-666
Sodium bis(fluorosulfonyl)imide 95%
Sodium bis(fluorosulfonyl)imide

Precaución: for Research Use Only

Formula: F₂NNaO₄S₂
MW: 203.11 g/mol
MDL Number: MFCD29905041
CAS Number: 100669-96-3
UN: 1759
ADR: 8 Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR