(4-Bromo-benzothiazol-2-yl)-carbamic acid tert-butyl ester 95%

Supplier: Ambeed
Warning

A759379-100MG A759379-1G A759379-250MG
76652-656EA 54.11 USD
76652-656 76652-658 76652-660
(4-Bromo-benzothiazol-2-yl)-carbamic acid tert-butyl ester 95%
(4-Bromo-benzothiazol-2-yl)-carbamic acid tert-butyl ester

Precaución: for Research Use Only

Formula: C₁₂H₁₃BrN₂O₂S
MW: 329.2128 g/mol
MDL Number: MFCD28154577
CAS Number: 1823565-33-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR