Fmoc-L-Dap-OtBu HCl 95%

Supplier: Ambeed
Warning

A759344-100MG A759344-250MG
76652-652EA 55.17 USD
76652-652 76652-654
Fmoc-L-Dap-OtBu HCl 95%
Fmoc-L-Dap-OtBu HCl

Precaución: for Research Use Only

Formula: C₂₂H₂₇ClN₂O₄
MW: 418.9138 g/mol
MDL Number: MFCD21363165
CAS Number: 2098497-06-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR