Di-tert-butyl butane-1,4-diylbis((3-aminopropyl)carbamate) 95%

Supplier: Ambeed
Warning

A759246-250MG A759246-1G
76652-650EA 141.1 USD
76652-650 77573-452
Di-tert-butyl butane-1,4-diylbis((3-aminopropyl)carbamate) 95%
Di-tert-butyl butane-1,4-diylbis((3-aminopropyl)carbamate)

Precaución: for Research Use Only

Formula: C₂₀H₄₂N₄O₄
MW: 402.5719 g/mol
MDL Number: MFCD08274632
CAS Number: 177213-61-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR