3-(5,5-Dimethyl-1,3,2-dioxaborinan-2-yl)aniline 95%

Supplier: Ambeed
Warning

A759016-1G A759016-250MG
76652-646EA 58 USD
76652-646 76652-648
3-(5,5-Dimethyl-1,3,2-dioxaborinan-2-yl)aniline 95%
3-(5,5-Dimethyl-1,3,2-dioxaborinan-2-yl)aniline

Precaución: for Research Use Only

Formula: C₁₁H₁₆BNO₂
MW: 205.0612 g/mol
MDL Number: MFCD24393354
CAS Number: 220080-93-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR