(4-(N-(2-((tert-Butoxycarbonyl)amino)ethyl)sulfamoyl)phenyl)boronic acid 96%

Supplier: Ambeed
Warning

A758852-100MG A758852-1G A758852-250MG
76652-640EA 37.14 USD
76652-640 76652-642 76652-644
(4-(N-(2-((tert-Butoxycarbonyl)amino)ethyl)sulfamoyl)phenyl)boronic acid 96%
(4-(N-(2-((tert-Butoxycarbonyl)amino)ethyl)sulfamoyl)phenyl)boronic acid

Precaución: for Research Use Only

Formula: C₁₃H₂₁BN₂O₆S
MW: 344.1916 g/mol
MDL Number: MFCD31543952
CAS Number: 874459-63-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR