[2,2'-Bis(4-tert-butylpyridine)]bis[2-(2,4-difluorophenyl)pyridine]iridium(III) hexafluorophosphate 97%

Supplier: Ambeed
Warning

A757574-100MG A757574-1G A757574-250MG A757574-5G
76652-628EA 40 USD
76652-628 76652-630 76652-632 77783-961
[2,2'-Bis(4-tert-butylpyridine)]bis[2-(2,4-difluorophenyl)pyridine]iridium(III) hexafluorophosphate 97%
[2,2'-Bis(4-tert-butylpyridine)]bis[2-(2,4-difluorophenyl)pyridine]iridium(III) hexafluorophosphate

Precaución: for Research Use Only

Formula: C₄₀H₃₆F₁₀IrN₄P
MW: 985.9200 g/mol
MDL Number: MFCD31657504
CAS Number: 1072067-44-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR