6-Fuoro-N,N-dimethylpyridin-3-amine 95%

Supplier: Ambeed
Warning

A757561-1G A757561-250MG
76652-624EA 197 USD
76652-624 76652-626
6-Fuoro-N,N-dimethylpyridin-3-amine 95%
6-Fuoro-N,N-dimethylpyridin-3-amine

Precaución: for Research Use Only

Formula: C₇H₉FN₂
MDL Number: MFCD28154575
CAS Number: 1823624-32-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR