6-(4-Isopropyl-4H-1,2,4-triazol-3-yl)pyridin-2-amine 95%

Supplier: Ambeed
Warning

A757107-250MG A757107-100MG
76652-610EA 43.5 USD
76652-610 77618-088
6-(4-Isopropyl-4H-1,2,4-triazol-3-yl)pyridin-2-amine 95%
6-(4-Isopropyl-4H-1,2,4-triazol-3-yl)pyridin-2-amine

Precaución: for Research Use Only

Formula: C₁₀H₁₃N₅
MW: 203.2437 g/mol
MDL Number: MFCD24127938
CAS Number: 1448427-99-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR