tert-Butyl (23-hydroxy-3,6,9,12,15,18,21-heptaoxatricosyl)carbamate 95%

Supplier: Ambeed
Warning

Synonyms: N-Boc-PEG8-alcohol

A756903-100MG A756903-1G A756903-250MG
76652-604EA 80.63 USD
76652-604 76652-606 76652-608
tert-Butyl (23-hydroxy-3,6,9,12,15,18,21-heptaoxatricosyl)carbamate 95%
tert-Butyl (23-hydroxy-3,6,9,12,15,18,21-heptaoxatricosyl)carbamate

Precaución: for Research Use Only

Formula: C₂₁H₄₃NO₁₀
MW: 469.5668 g/mol
MDL Number: MFCD28976659
CAS Number: 1345337-22-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR