1-(5-(Aminomethyl)-2-nitrophenyl)ethanol 95%

Supplier: Ambeed
Warning

A756593-100MG A756593-1G A756593-250MG
76652-598EA 103.97 USD
76652-598 76652-600 76652-602
1-(5-(Aminomethyl)-2-nitrophenyl)ethanol 95%
1-(5-(Aminomethyl)-2-nitrophenyl)ethanol

Precaución: for Research Use Only

Formula: C₉H₁₂N₂O₃
MW: 196.2032 g/mol
MDL Number: MFCD28976671
CAS Number: 138957-91-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR