Methyl 6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)chroman-2-carboxylate 97%

Supplier: Ambeed
Warning

A756271-100MG A756271-1G A756271-250MG A756271-5G
76652-586EA 37.14 USD
76652-586 76652-588 76652-590 76652-592
Methyl 6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)chroman-2-carboxylate 97%
Methyl 6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)chroman-2-carboxylate

Precaución: for Research Use Only

Formula: C₁₇H₂₃BO₅
MW: 318.1725 g/mol
MDL Number: MFCD30474681
CAS Number: 2152654-65-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR